(S)-(+)-Methyl-3-hydroxybutyrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl (S)-(+)-3-Hydroxybutyrate, (S)-Methyl 3-hydroxybutanoate, Methyl (S)-(+)-3-hydroxybutyrate

L19996-06 L19996-14
CAAAAL19996-03EA 108.02 CAD
CAAAAL19996-03 CAAAAL19996-06 CAAAAL19996-14
(S)-(+)-Methyl-3-hydroxybutyrate 98%
(S)-(+)-Methyl 3-hydroxybutyrate

Purity: 98% CAS No.: 53562-86-0 Molecular Formula: C5H10O3 Formula Weight: 118.13 Flash Point: 73°(163°F) Refractive Index: 1.4220 Boiling Point: 56-58°/11mm Beilstein Reference: 1720583 Optical Rotation: +48.5° (c=1.3 in chloroform) MDL No.: MFCD00064461

Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 56…58 °C/11 mm
Density: 1.055
Flash Pt: 73 °C(163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064461
CAS Number: 53562-86-0
EINECS: 258-628-8

Order Now

Specification Test Results

Optical rotation +48.5° (c=1.3 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR