tert-Butyl isocyanide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19747-03
L19747-06
CAAAAL19747-03EA
83.9
CAD
CAAAAL19747-03
CAAAAL19747-06
tert-Butyl isocyanide 98%
tert-Butyl isocyanide
Fieser: 2,50 9,82 11,99 12,95 14,63 20,62 21,77
Purity: 98%
CAS No.: 7188-38-7
Molecular Formula: C5H9N
Formula Weight: 83.13
Storage Temperature: -30°C to -10°C
Flash Point: -2°(28°F)
Refractive Index: 1.3755
Odor: Stench
Boiling Point: 90-92°
Beilstein Reference: 1903156
UN No.: 1992
MDL No.: MFCD00000002
Formula:
C₅H₉N Boiling Pt: 90…92 °C Density: 0.735 Flash Pt: -2 °C(28 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000002 CAS Number: 7188-38-7 EINECS: 230-553-5 UN: 1992 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...