Methyl-3-methylcrotonate 98%

Supplier: Thermo Scientific Chemicals
Warning

L19731-09 L19731-18
CAAAAL19731-09EA 121.5 CAD
CAAAAL19731-09 CAAAAL19731-18
Methyl-3-methylcrotonate 98%
Methyl 3-methylcrotonate

Purity: 98% CAS No.: 924-50-5 Molecular Formula: C6H10O2 Formula Weight: 114.14 Flash Point: 33°(91°F) Refractive Index: 1.4375 Melting Point: -41° Boiling Point: 134-139° Beilstein Reference: 1741592 UN No.: 3272 MDL No.: MFCD00043940

Formula: C₆H₁₀O₂
Boiling Pt: 134…139 °C
Melting Pt: –41 °C
Density: 0.936
Flash Pt: 33 °C(91 °F)
Storage Temperature: Ambient
MDL Number: MFCD00043940
CAS Number: 924-50-5
EINECS: 213-107-4
UN: 3272
ADR: "3",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR