Bismuth (III) trifluoromethanesulfonate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Bismuth(III) trifluoromethanesulfonate, Bismuth triflate, Bismuth(III) trifluoromethanesulphonate

L19687-06 L19687-14
CAAAAL19687-06EA 114.08 CAD
CAAAAL19687-06 CAAAAL19687-14
Bismuth (III) trifluoromethanesulfonate 99%
Bismuth (III) trifluoromethanesulphonate

Fieser: 21,59


Purity: 99% CAS No.: 88189-03-1 Molecular Formula: C3BiF9O9S3 Formula Weight: 656.19 Melting Point: 300° UN No.: 3261 MDL No.: MFCD02093669

Formula: C₃BiF₉O₉S₃
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD02093669
CAS Number: 88189-03-1
UN: 3261
ADR: "8",III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR