Lanthanum(III) trifluoromethanesulfonate, anhydrous 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Lanthanum(III) trifluoromethanesulphonate, Lanthanum trifluoromethanesulphonate, Lanthanum trifluoromethanesulfonate, Lanthanum(III) trifluoromethanesulfonate, Lanthanum triflate
L19657-06
L19657-14
CAAAAL19657-06EA
73.31
CAD
CAAAAL19657-06
CAAAAL19657-14
Lanthanum(III) trifluoromethanesulfonate, anhydrous 99%
Lanthanum (III) trifluoromethanesulphonate
Purity: 99%
CAS No.: 52093-26-2
Molecular Formula: C3F9LaO9S3
Formula Weight: 586.10
Beilstein Reference: 9009103
UN No.: 3261
MDL No.: MFCD00077560
Formula:
C₃F₉LaO₉S₃ MW: 586.12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00077560 CAS Number: 52093-26-2 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...