(R)-(-)-2-Chloromandelic acid ≥99%, ee 99%, ChiPros®

Supplier: Thermo Scientific Chemicals
Warning

L19586-03 L19586-06 L19586-14
CAAAAL19586-03EA 52.3 CAD
CAAAAL19586-03 CAAAAL19586-06 CAAAAL19586-14
(R)-(-)-2-Chloromandelic acid ≥99%, ee 99%, ChiPros®
(R)-(-)-2-Chloromandelic acid

Purity: 99+% CAS No.: 52950-18-2 Molecular Formula: C8H7ClO3 Formula Weight: 186.60 Melting Point: 119-121° Beilstein Reference: 2693370

Formula: C₈H₇ClO₃
Melting Pt: 119…121 °C
Density: 1.48
Storage Temperature: Ambient
MDL Number: MFCD00798429
CAS Number: 52950-18-2
EINECS: 444-040-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR