(R)-(+)-Hydroxypyrrolidine 99%, ee 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19499-03
L19499-06
CAAAAL19499-03EA
106.33
CAD
CAAAAL19499-03
CAAAAL19499-06
(R)-(+)-Hydroxypyrrolidine 99%, ee 99%
(R)-(+)-Hydroxypyrrolidine
Purity: 99%
CAS No.: 2799-21-5
Molecular Formula: C4H9NO
Formula Weight: 87.12
Flash Point:
Formula:
C₄H₉NO Boiling Pt: 108…110 °C/8 mm Melting Pt: 15 °C Density: 1.048 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00145220 CAS Number: 2799-21-5 |
Specification Test Results
Optical rotation | +6.5° (c=3.5 in methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...