(S)-(-)-Hydroxypyrrolidine 99%, ee 99%

Supplier: Thermo Scientific Chemicals
Warning

L19498-03 L19498-06 L19498-MD
CAAAAL19498-03EA 268.6 CAD
CAAAAL19498-03 CAAAAL19498-06 CAAAAL19498-MD
(S)-(-)-Hydroxypyrrolidine 99%, ee 99%
(S)-(-)-Hydroxypyrrolidine

Purity: 99% CAS No.: 100243-39-8 Molecular Formula: C4H9NO Formula Weight: 87.12 Flash Point: 110°(230°F) Refractive Index: 1.4900 Melting Point: 15° Boiling Point: 108-110°/8mm Beilstein Reference: 4652606 Optical Rotation: -6.5° (c=3.5 in methanol) MDL No.: MFCD00192426

Formula: C₄H₉NO
Boiling Pt: 108…110 °C/8 mm
Melting Pt: 15 °C
Density: 1.048
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00192426
CAS Number: 100243-39-8

Order Now

Specification Test Results

Optical rotation -6.5° (c=3.5 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR