(R)-(+)-Tetrahydro-2-furoic acid ≥98%, ee 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(R)-2-Tetrahydrofuroic acid
L19410-03
L19410-06
L19410-14
CAAAAL19410-03EA
62.41
CAD
CAAAAL19410-03
CAAAAL19410-06
CAAAAL19410-14
(R)-(+)-Tetrahydro-2-furoic acid ≥98%, ee 98%
(R)-(+)-Tetrahydro-2-furoic acid
Purity: 98+%
CAS No.: 87392-05-0
Molecular Formula: C5H8O3
Formula Weight: 116.12
Flash Point: 139°(282°F)
Refractive Index: 1.4600
Boiling Point: 244°
Beilstein Reference: 4658739
Optical Rotation: +4° (c=1 in methanol)
UN No.: 3265
MDL No.: MFCD00211271
Formula:
C₅H₈O₃ MW: 116.12 g/mol Boiling Pt: 244 °C Density: 1.213 Flash Pt: 139 °C(282 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00211271 CAS Number: 87392-05-0 UN: 3265 ADR: "8",III |
Specification Test Results
Optical rotation | +4° (c=1 in methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...