(R)-(+)-4-Chloro-α-methylbenzylamine 97%, ee 98%, ChiPros®
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19119-03
L19119-06
CAAAAL19119-03EA
69.05
CAD
CAAAAL19119-03
CAAAAL19119-06
CAAAAL19119-14
(R)-(+)-4-Chloro-α-methylbenzylamine 97%, ee 98%, ChiPros®
(R)-(+)-4-Chloro-α-methylbenzylamine
Purity: 97%
CAS No.: 27298-99-3
Molecular Formula: C8H10ClN
Formula Weight: 155.63
Flash Point: 115°(239°F)
Refractive Index: 1.5418
Boiling Point: 232°
Beilstein Reference: 2412317
Optical Rotation: +31° (neat)
UN No.: 2922
MDL No.: MFCD00671639
Formula:
C₈H₁₀ClN Boiling Pt: 232 °C Density: 1.110 Flash Pt: 115 °C(239 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00671639 CAS Number: 27298-99-3 UN: 2810 ADR: "6.1",III |
Specification Test Results
Optical rotation | +31° (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...