Tetrabutylammonium dihydrogentrifluoride 50 - 55% (w/w) in 1,2-dichloroethane
Supplier: Thermo Scientific Chemicals
|
Danger
|
L17891-03
L17891-06
CAAAAL17891-03EA
46.93
CAD
CAAAAL17891-03
CAAAAL17891-06
Tetrabutylammonium dihydrogentrifluoride 50 - 55% (w/w) in 1,2-dichloroethane
Tetrabutylammonium dihydrogentrifluoride
Fieser: 20,357
CAS No.: 99337-56-1
Molecular Formula: [CH3(CH2)3]4N(H2F3)
Formula Weight: 301.48
Flash Point: 13°(55°F)
UN No.: 2924
MDL No.: MFCD00145365
Formula:
C₁₆H₃₈F₃N Flash Pt: 13 °C (55 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00145365 CAS Number: 99337-56-1 UN: 2924 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...