3-Chlor-1,2-benzisothiazol 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Chlorobenzo[d]isothiazole, 3-Chloro-1,2-benzisothiazole

L17791-06 L17791-14
CAAAAL17791-06EA 113.13 CAD
CAAAAL17791-06 CAAAAL17791-14
3-Chlor-1,2-benzisothiazol 97+%
3-Chlor-1,2-benzisothiazol

Purity: 99% CAS No.: 7716-66-7 Molecular Formula: C7H4ClNS Formula Weight: 169.63 Melting Point: 37-44° Boiling Point: 80-86°/0.75mm MDL No.: MFCD00673254

Formula: C₇H₄ClNS
Boiling Pt: 80…86 °C/0.75 mm
Melting Pt: 37…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00673254
CAS Number: 7716-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR