4-Iodobenzaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Warning

L17448-03 L17448-06 L17448-MD
CAAAAL17448-03EA 157.68 CAD
CAAAAL17448-03 CAAAAL17448-06 CAAAAL17448-MD
4-Iodobenzaldehyde 98+%
4-Iodobenzaldehyde

Purity: 98+% CAS No.: 15164-44-0 Molecular Formula: C7H5IO Formula Weight: 232.02 Storage Temperature: 2°C to 8°C Melting Point: 76-78° Beilstein Reference: 636101 MDL No.: MFCD00039576

Formula: C₇H₅IO
MW: 232.02 g/mol
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00039576
CAS Number: 15164-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR