1,1,1-Trifluoro-2-butanone 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L16868-03
L16868-06
CAAAAL16868-03EA
62.73
CAD
CAAAAL16868-03
CAAAAL16868-06
1,1,1-Trifluoro-2-butanone 96%
1,1,1-Trifluoro-2-butanone
Purity: 96%
CAS No.: 381-88-4
Molecular Formula: C4H5F3O
Formula Weight: 126.08
Flash Point: -17°(1°F)
Refractive Index: 1.3070
Boiling Point: 44-45°
Beilstein Reference: 1702220
UN No.: 1224
MDL No.: MFCD00190641
Formula:
C₄H₅F₃O Boiling Pt: 44…45 °C Density: 0.929 Flash Pt: -17 °C(1 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00190641 CAS Number: 381-88-4 UN: 1224 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...