5-Fluoro-2'-deoxyuridine (floxuridine) 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
5-Fluoro-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Fluoro-2'-deoxyuridine, Floxuridine
L16497-MC
L16497-ME
CAAAAL16497-MCEA
184.86
CAD
CAAAAL16497-MC
CAAAAL16497-ME
5-Fluoro-2'-deoxyuridine (floxuridine) 98+%
5-Fluoro-2'-deoxyuridine (Floxuridine)
Purity: 98+%
CAS No.: 50-91-9
Molecular Formula: C9H11FN2O5
Formula Weight: 246.20
Melting Point: 148-151°
Beilstein Reference: 2645818
Merck Reference: 14,4112
UN No.: 2811
MDL No.: MFCD00006530
Formula:
C₉H₁₁FN₂O₅ Melting Pt: 148…151 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006530 CAS Number: 50-91-9 EINECS: 200-072-5 UN: 2811 ADR: "6.1",III Merck Index: 14,4112 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...