(3-Acryloxypropyl)trimethoxysilane 94% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-(Trimethoxysilyl)propyl acrylate
Stabilization:
100 ppm Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
L16400-06
L16400-14
CAAAAL16400-06EA
139.99
CAD
CAAAAL16400-06
CAAAAL16400-14
(3-Acryloxypropyl)trimethoxysilane 94% stabilized
(3-Acryloxypropyl)trimethoxysilane
Purity: 94%
CAS No.: 4369-14-6
Molecular Formula: C9H18O5Si
Formula Weight: 234.33
Flash Point: 123°(253°F)
Refractive Index: 1.4290
Boiling Point: 95-100°/3mm
Beilstein Reference: 2046320
UN No.: 1760
MDL No.: MFCD00054803
Formula:
C₉H₁₈O₅Si MW: 234.32 g/mol Boiling Pt: 95…100 °C/3 mm Density: 1.059 Flash Pt: 123 °C (253 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00054803 CAS Number: 4369-14-6 EINECS: 419-560-6 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...