(S)-(-)-3-Methoxy-α-methylbenzylamine ≥99%, ee 99%, ChiPros®
Supplier: Thermo Scientific Chemicals
|
Danger
|
L16324-03
L16324-06
L16324-14
CAAAAL16324-03EA
79
CAD
CAAAAL16324-03
CAAAAL16324-06
CAAAAL16324-14
(S)-(-)-3-Methoxy-α-methylbenzylamine ≥99%, ee 99%, ChiPros®
(S)-(-)-3-Methoxy-α-methylbenzylamine
Purity: 99+%
CAS No.: 82796-69-8
Molecular Formula: C9H13NO
Formula Weight: 151.21
Flash Point:
Formula:
C₉H₁₃NO Boiling Pt: 66 °C/0.38 mm Melting Pt: 5 °C Density: 1.023 Flash Pt: >100 °C(212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00671656 CAS Number: 82796-69-8 EINECS: 632-902-0 UN: 2735 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...