(R)-(+)-3-Methoxy-α-methylbenzylamine ≥99%, ee 98%, ChiPros®
Supplier: Thermo Scientific Chemicals
|
Danger
|
L16323-06
L16323-14
CAAAAL16323-06EA
225.94
CAD
CAAAAL16323-06
CAAAAL16323-14
(R)-(+)-3-Methoxy-α-methylbenzylamine ≥99%, ee 98%, ChiPros®
(R)-(+)-3-Methoxy-α-methylbenzylamine
Purity: 99+%
CAS No.: 88196-70-7
Molecular Formula: C9H13NO
Formula Weight: 151.21
Flash Point:
Formula:
C₉H₁₃NO Boiling Pt: 66 °C/0.38 mm Melting Pt: 5 °C Density: 1.023 Flash Pt: >100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00671655 CAS Number: 88196-70-7 EINECS: 618-126-5 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...