(S)-(-)-4-Methoxy-α-methylbenzylamine ≥99%, ee 98%, ChiPros®

Supplier: Thermo Scientific Chemicals
Danger

L16322-03 L16322-06 L16322-14
CAAAAL16322-03EA 60.83 CAD
CAAAAL16322-03 CAAAAL16322-06 CAAAAL16322-14
(S)-(-)-4-Methoxy-α-methylbenzylamine ≥99%, ee 98%, ChiPros®
(S)-(-)-4-Methoxy-α-methylbenzylamine

Purity: 99+% CAS No.: 41851-59-6 Molecular Formula: C9H13NO Formula Weight: 151.21 Refractive Index: 1.5330 Melting Point: -20° Boiling Point: 65°/0.38mm Beilstein Reference: 3196456 Optical Rotation: -34° (neat) UN No.: 2735 MDL No.: MFCD00671660

Formula: C₉H₁₃NO
Boiling Pt: 65 °C/0.38 mm
Melting Pt: < –20 °C
Density: 1.020
Storage Temperature: Ambient
MDL Number: MFCD00671660
CAS Number: 41851-59-6
EINECS: 609-962-1
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Optical rotation -34° (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR