(R)-(+)-α-Ethylbenzylamine ≥99%, ee 98%, ChiPros®
Supplier: Thermo Scientific Chemicals
|
Danger
|
L16319-06
L16319-14
CAAAAL16319-06EA
115.5
CAD
CAAAAL16319-06
CAAAAL16319-14
(R)-(+)-α-Ethylbenzylamine ≥99%, ee 98%, ChiPros®
(R)-(+)-α-Ethylbenzylamine
For synthesis of optically active products
Purity: 99+%
CAS No.: 3082-64-2
Molecular Formula: C9H13N
Formula Weight: 135.21
Flash Point: 77°(170°F)
Refractive Index: 1.5200
Odor: Amine-like
Melting Point: -69°
Boiling Point: 205°
Beilstein Reference: 3029904
Optical Rotation: +20° (neat)
UN No.: 2735
MDL No.: MFCD00083057
Formula:
C₉H₁₃N Boiling Pt: 205 °C Melting Pt: –69 °C Density: 0.930 Flash Pt: 77 °C(170 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00083057 CAS Number: 3082-64-2 EINECS: 608-544-6 UN: 2735 ADR: "8",III |
Specification Test Results
Optical rotation | +20° (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...