Glycine-tert-butyl ester(H-Gly-OtBu) 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
tert-Butyl 2-aminoacetate, tert-Butyl Glycinate
L16258-03
L16258-06
CAAAAL16258-03EA
124.19
CAD
CAAAAL16258-03
CAAAAL16258-06
Glycine-tert-butyl ester(H-Gly-OtBu) 97%
Glycine tert-butyl ester (H-Gly-OtBu)
Purity: 97%
CAS No.: 6456-74-2
Molecular Formula: H2NCH2CO2C(CH3)3
Formula Weight: 131.18
Storage Temperature: 2°C to 8°C
Refractive Index: 1.4240
Boiling Point: 29-31°/2mm
Beilstein Reference: 2039000
MDL No.: MFCD00038194
Formula:
C₆H₁₃NO₂ MW: 131.18 g/mol Boiling Pt: 29…31 °C/2 mm Density: 0.960 Storage Temperature: Ambient |
MDL Number:
MFCD00038194 CAS Number: 6456-74-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...