Allylboronic acid pinacol ester 98+%

Supplier: Thermo Scientific Chemicals
Warning

L16232-03 L16232-06
CAAAAL16232-03EA 69.99 CAD
CAAAAL16232-03 CAAAAL16232-06
Allylboronic acid pinacol ester 98+%
Allylboronic acid pinacol ester

Purity: 98+% CAS No.: 72824-04-5 Molecular Formula: C9H17BO2 Formula Weight: 168.05 Flash Point: 46°(140°F) Refractive Index: 1.4270 Boiling Point: 50-53°/5mm Beilstein Reference: 4244068 UN No.: 3272 MDL No.: MFCD00013347

Formula: C₉H₁₇BO₂
Boiling Pt: 50…53 °C/5 mm
Density: 0.890
Flash Pt: 46 °C(140 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013347
CAS Number: 72824-04-5
UN: 1993
ADR: "3",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR