tert-Butyl nitrite tech. 90%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
L16135-14
L16135-22
CAAAAL16135-14EA
59.09
CAD
CAAAAL16135-14
CAAAAL16135-22
tert-Butyl nitrite tech. 90%, Technical Grade
tert-Butyl nitrite
Fieser: 1,97 7,48
Purity: 90%
CAS No.: 540-80-7
Molecular Formula: (CH3)3CONO
Formula Weight: 103.12
Storage Temperature: 2°C to 8°C
Flash Point: -23°(-9°F)
Refractive Index: 1.3690
Odor: Sweet
Boiling Point: 61-63°
Beilstein Reference: 1209339
Merck Reference: 14,1583
UN No.: 2351
MDL No.: MFCD00002055
Formula:
C₄H₉NO₂ Boiling Pt: 61…63 °C Density: 0.867 Flash Pt: –23 °C (–9 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002055 CAS Number: 540-80-7 EINECS: 208-757-0 UN: 2351 ADR: 3,III Merck Index: 14,1583 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...