Zinc trifluoromethanesulfonate 98%

Supplier: Thermo Scientific Chemicals
Danger

L15969-06 L15969-14
CAAAAL15969-06EA 85.48 CAD
CAAAAL15969-06 CAAAAL15969-14
Zinc trifluoromethanesulfonate 98%
Zinc trifluoromethanesulphonate

Fieser: 12,577 16,392 18,412 21,494


Purity: 98% CAS No.: 54010-75-2 Molecular Formula: C2F6O6S2Zn Formula Weight: 363.51 Melting Point: 300° Beilstein Reference: 4028195 UN No.: 3261 MDL No.: MFCD00013229

Formula: C₂F₆O₆S₂Zn
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00013229
CAS Number: 54010-75-2
EINECS: 258-922-6
UN: 3261
ADR: "8",III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR