N-Benzylglycine ethyl ester 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L15496-14
L15496-22
L15496-36
CAAAAL15496-14EA
104.91
CAD
CAAAAL15496-14
CAAAAL15496-22
CAAAAL15496-36
N-Benzylglycine ethyl ester 97%
N-Benzylglycine ethyl ester
Purity: 97%
CAS No.: 6436-90-4
Molecular Formula: C6H5CH2NHCH2CO2CH2CH3
Formula Weight: 193.25
Storage Temperature: 2°C to 8°C
Flash Point: 112°(233°F)
Refractive Index: 1.5053
Boiling Point: 102-104°/1mm
Beilstein Reference: 2104816
MDL No.: MFCD00009174
Formula:
C₁₁H₁₅NO₂ Boiling Pt: 102…104 °C/1 mm Density: 1.048 Flash Pt: 112 °C(233 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009174 CAS Number: 6436-90-4 EINECS: 229-218-6 UN: 2735 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...