D-(-)-Quinic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

L15238-06 L15238-14
CAAAAL15238-06EA 54.04 CAD
CAAAAL15238-06 CAAAAL15238-14
D-(-)-Quinic acid 98%
D(-)-Quinic acid

Purity: 98% CAS No.: 77-95-2 Molecular Formula: C7H12O6 Formula Weight: 192.17 Odor: Odorless Melting Point: 162-167° Beilstein Reference: 2212412 Merck Reference: 14,8059 Optical Rotation: -44° (c=20 in water) MDL No.: MFCD00003864

Formula: C₇H₁₂O₆
Melting Pt: 162…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00003864
CAS Number: 77-95-2
EINECS: 201-072-8
Merck Index: 14,8059

Order Now

Specification Test Results

Optical rotation -44° (c=20 in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR