Bis(cyclopentadienyl)zirconium hydridochloride 95%

Supplier: Thermo Scientific Chemicals
Danger

L15164-03 L15164-06 L15164-14
CAAAAL15164-03EA 61.3 CAD
CAAAAL15164-03 CAAAAL15164-06 CAAAAL15164-14
Bis(cyclopentadienyl)zirconium hydridochloride 95%
Bis(cyclopentadienyl)zirconium hydridochloride

Fieser: 6,175 14,81 15,80 16,72 17,33 18,416 19,415 20,445 21,497


Purity: 95% CAS No.: 37342-97-5 Molecular Formula: C10H11ClZr Formula Weight: 257.87 Storage Temperature: 2°C to 8°C Melting Point: 300° UN No.: 3131 MDL No.: MFCD02089401

Formula: C₁₀H₁₁ClZr
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD02089401
CAS Number: 37342-97-5
EINECS: 253-479-5
UN: 3131
ADR: 4.3,III
Merck Index: 14,8396

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR