Valeramide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Pentanamide
L14601-06
L14601-14
CAAAAL14601-06EA
59.88
CAD
CAAAAL14601-06
CAAAAL14601-14
Valeramide 97%
Valeramide
Purity: 97%
CAS No.: 626-97-1
Molecular Formula: CH3(CH2)3CONH2
Formula Weight: 101.15
Melting Point: 102-104°
Beilstein Reference: 1740792
Formula:
C₅H₁₀O₂ MW: 102.13 g/mol Melting Pt: 102…104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041895 CAS Number: 626-97-1 EINECS: 210-974-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...