Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate, (max. 10% H₂O) 99%

Supplier: Thermo Scientific Chemicals

L14522-03 L14522-06
CAAAAL14522-03EA 75.37 CAD
CAAAAL14522-03 CAAAAL14522-06
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate, (max. 10% H₂O) 99%
Adenosine 5'-triphosphate disodium salt (ATP disodium salt) hydrate

Purity: 99% CAS No.: 34369-07-8 Molecular Formula: C10H14N5Na2O13P3.xH2O Formula Weight: 551.15(anhy) Storage Temperature: -30°C to -10°C Merck Reference: 14,155 Optical Rotation: -19° (c=3 in water) MDL No.: MFCD00150755

Formula: C₁₀H₁₄N₅Na₂O₁₃P₃·xH₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00150755
CAS Number: 34369-07-8
EINECS: 213-579-1
Merck Index: 14,155

Order Now

Specification Test Results

Optical rotation -19° (c=3 in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR