4-tert-Butylcalix[6]arene 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
p-tert-Butylcalix[6]arene
L14473-03
L14473-06
L14473-14
CAAAAL14473-03EA
49.3
CAD
CAAAAL14473-03
CAAAAL14473-06
CAAAAL14473-14
4-tert-Butylcalix[6]arene 96%
4-tert-Butylcalix[6]arene
Purity: 96%
CAS No.: 78092-53-2
Molecular Formula: C66H84O6
Formula Weight: 973.40
Melting Point: 377-381°
Beilstein Reference: 2611918
Formula:
C₆₆H₈₄O₆ MW: 973.39 g/mol Melting Pt: 377…381 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075464 CAS Number: 78092-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...