Sodium mercaptoacetate (sodium thioglycolate) 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L14356-14
L14356-22
L14356-36
CAAAAL14356-14EA
49.61
CAD
CAAAAL14356-14
CAAAAL14356-22
CAAAAL14356-36
Sodium mercaptoacetate (sodium thioglycolate) 97%
Sodium mercaptoacetate (Sodium thioglycolate)
Reagent that protects tryptophan in amino acid analysis
Purity: 97%
CAS No.: 367-51-1
Molecular Formula: HSCH2CO2Na
Formula Weight: 114.10
Odor: Stench
pH: ~6
Melting Point: ca 305°
Beilstein Reference: 4569109
Merck Reference: 14,8692
UN No.: 3335
MDL No.: MFCD00043386
Formula:
HSCH₂COONa Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043386 CAS Number: 367-51-1 EINECS: 206-696-4 UN: 2923 ADR: "8",III Merck Index: 14,8692 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...