Benzyloxyacetyl chloride 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L14337-06
L14337-14
CAAAAL14337-03EA
54.08
CAD
CAAAAL14337-03
CAAAAL14337-06
CAAAAL14337-14
Benzyloxyacetyl chloride 95%
Benzyloxyacetyl chloride
Purity: 95%
CAS No.: 19810-31-2
Molecular Formula: C9H9ClO2
Formula Weight: 184.62
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₉H₉ClO₂ Boiling Pt: 84…87 °C/0.4 mm Density: 1.189 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010768 CAS Number: 19810-31-2 UN: 3265 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...