N-Acetyl-L-proline 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Acetyl-L-Proline, Ac-Pro-OH
L14300-03
L14300-06
CAAAAL14300-03EA
56.88
CAD
CAAAAL14300-03
CAAAAL14300-06
N-Acetyl-L-proline 99%
N-Acetyl-L-proline
Purity: 99%
CAS No.: 68-95-1
Molecular Formula: C7H11NO3
Formula Weight: 157.18
Storage Temperature: 2°C to 8°C
Melting Point: 117-118°
Beilstein Reference: 83200
Optical Rotation: -116° (c=1 in water)
MDL No.: MFCD00020837
Formula:
C₇H₁₁NO₃ MW: 157.169 g/mol Melting Pt: 117…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020837 CAS Number: 68-95-1 EINECS: 200-698-9 |
Specification Test Results
Optical rotation | -116° (c=1 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...