2'-Deoxycytidine hydrochloride 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one hydrochloride

L14153-03 L14153-MD
CAAAAL14153-03EA 99.38 CAD
CAAAAL14153-03 CAAAAL14153-MD
2'-Deoxycytidine hydrochloride 99%
2'-Deoxycytidine hydrochloride

Purity: 99% CAS No.: 3992-42-5 Molecular Formula: C9H13N3O4.HCl Formula Weight: 263.68 Odor: Odorless Melting Point: ca 170° dec. Beilstein Reference: 3576124 Optical Rotation: +57° (c=1 in water) MDL No.: MFCD00012840

Formula: C₉H₁₄ClN₃O₄
MW: 263.68 g/mol
Melting Pt: ca 170 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012840
CAS Number: 3992-42-5
EINECS: 223-639-9

Order Now

Specification Test Results

Optical rotation +57° (c=1 in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR