(R)-(-)-Methyl-3-hydroxybutyrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-Methyl 3-hydroxybutanoate, Methyl (R)-3-hydroxybutyrate, Methyl (R)-(-)-3-Hydroxybutyrate

L14151-03 L14151-06 L14151-14
CAAAAL14151-03EA 64.78 CAD
CAAAAL14151-03 CAAAAL14151-06 CAAAAL14151-14
(R)-(-)-Methyl-3-hydroxybutyrate 98%
(R)-(-)-Methyl 3-hydroxybutyrate

Fieser: 15,207 16,218


Purity: 98% CAS No.: 3976-69-0 Molecular Formula: C5H10O3 Formula Weight: 118.13 Flash Point: 73°(163°F) Refractive Index: 1.4220 Odor: Pleasant Boiling Point: 56-58°/11mm Beilstein Reference: 3648161 Optical Rotation: -48.5° (c=1.3 in chloroform) MDL No.: MFCD00063289

Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 56…58 °C/11 mm
Density: 1.055
Flash Pt: 73 °C(163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00063289
CAS Number: 3976-69-0
EINECS: 223-610-0

Order Now

Specification Test Results

Optical rotation -48.5° (c=1.3 in methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR