Methyl-3-bromopropionate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 3-bromopropanoate
L14127-14
L14127-22
CAAAAL14127-14EA
107.28
CAD
CAAAAL14127-14
CAAAAL14127-22
Methyl-3-bromopropionate 97%
Methyl 3-bromopropionate
Purity: 97%
CAS No.: 3395-91-3
Molecular Formula: Br(CH2)2CO2CH3
Formula Weight: 167.01
Flash Point: 75°(167°F)
Refractive Index: 1.4580
Boiling Point: 58-60°/12mm
Beilstein Reference: 1742949
MDL No.: MFCD00000250
Formula:
C₄H₇BrO₂ MW: 167 g/mol Boiling Pt: 58…60 °C/12 mm Density: 1.530 Flash Pt: 75 °C(167 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000250 CAS Number: 3395-91-3 EINECS: 222-247-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...