Adenosine-5'-monophosphoric acid (AMP), (max. 6% H₂O) 99% (dry weight)

Supplier: Thermo Scientific Chemicals

Synonyms: ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogen phosphate, 5'-Adenylic acid, Adenosine 5'-monophosphate, Adenosine monophosphate, 5'-AMP

L14051-03 L14051-06
CAAAAL14051-03EA 39.66 CAD
CAAAAL14051-03 CAAAAL14051-06
Adenosine-5'-monophosphoric acid (AMP), (max. 6% H₂O) 99% (dry weight)
Adenosine-5'-monophosphoric acid (AMP)

a useful ligand determinant that facilitate the binding of APS reductase inhibitors and activates adenosine receptor agonists.


5'-AMP is an activator of a class of protein kinases known as AMP-activated protein kinase, an activator of phosphorylase b in the polymerization reaction for the synthesis of branched polysaccharides, a reagent in the synthesis of adenosine-5?-phosphoimidazolide, an inhibitor of endogenous NMN adenylyltransferase (NMNAT) activity that converts NMN to NAD+.

Formula: C₁₀H₁₄N₅O₇P
MW: 347.22 g/mol
Melting Pt: ca 190 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005750
CAS Number: 61-19-8
EINECS: 200-500-0
Merck Index: 14,158

Order Now

Specification Test Results

Optical rotation -47° (c=2 in 2% NaOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR