Dimethoxymethylvinylsilane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L14035-14
L14035-22
CAAAAL14035-06EA
49.03
CAD
CAAAAL14035-06
CAAAAL14035-14
CAAAAL14035-22
Dimethoxymethylvinylsilane 97%
Dimethoxymethylvinylsilane
Purity: 97%
CAS No.: 16753-62-1
Molecular Formula: C5H12O2Si
Formula Weight: 132.23
Flash Point: 5°(41°F)
Refractive Index: 1.3950
Boiling Point: 102-104°
Beilstein Reference: 3043765
UN No.: 1993
MDL No.: MFCD00129170
Formula:
C₅H₁₂O₂Si Boiling Pt: 102…104 °C Density: 0.889 Flash Pt: 5 °C(41 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00129170 CAS Number: 16753-62-1 EINECS: 240-816-6 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...