Adenosine 5'-diphosphate disodium salt (ADP disodium salt), (max. 7% H₂O) 96% (dry weight)
Supplier: Thermo Scientific Chemicals
Synonyms:
Adenosine 5'-Diphosphate Disodium Salt, Sodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl hydrogen diphosphate
L14029-03
L14029-MD
CAAAAL14029-03EA
122.29
CAD
CAAAAL14029-03
CAAAAL14029-MD
Adenosine 5'-diphosphate disodium salt (ADP disodium salt), (max. 7% H₂O) 96% (dry weight)
Adenosine 5'-diphosphate disodium salt (ADP disodium salt)
Purity: 97%
CAS No.: 16178-48-6
Molecular Formula: C10H13N5Na2O10P2
Formula Weight: 471.17
Storage Temperature: -30°C to -10°C
MDL No.: MFCD00066635
Formula:
C₁₀H₁₃N₅Na₂O₁₀P₂ MW: 471.167 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00066635 CAS Number: 16178-48-6 EINECS: 240-314-7 |
Specification Test Results
Optical rotation | -30° (c=0.5 in 0.5M Na‚HPO„) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...