2,2-Bis(hydroxymethyl)propionic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid
L14014-22
L14014-36
CAAAAL14014-22EA
61.15
CAD
CAAAAL14014-22
CAAAAL14014-36
2,2-Bis(hydroxymethyl)propionic acid 98+%
2,2-Bis(hydroxymethyl)propionic acid
Purity: 98+%
CAS No.: 4767-03-7
Molecular Formula: C5H10O4
Formula Weight: 134.13
Melting Point: 187-191°
Beilstein Reference: 1756385
Merck Reference: 14,3253
MDL No.: MFCD00004199
Formula:
C₅H₁₀O₄ MW: 134.13 g/mol Melting Pt: 187…191 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004199 CAS Number: 4767-03-7 EINECS: 225-306-3 Merck Index: 14,3253 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...