Hydrogen peroxide-urea 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13940-14
L13940-22
L13940-36
CAAAAL13940-14EA
50.88
CAD
CAAAAL13940-14
CAAAAL13940-22
CAAAAL13940-36
Hydrogen peroxide-urea 97%
Hydrogen peroxide-urea
Fieser: 16,379 17,401 18,185 21,483
Purity: 97%
CAS No.: 124-43-6
Molecular Formula: CH4N2O.H2O2
Formula Weight: 94.07
Odor: Odorless
Melting Point: ca 90° dec.
Beilstein Reference: 3680414
Merck Reference: 14,1782
UN No.: 1511
MDL No.: MFCD00013119
Formula:
CH₆N₂O₃ Melting Pt: ca 90 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00013119 CAS Number: 124-43-6 EINECS: 204-701-4 UN: 1511 ADR: "5.1",III Merck Index: 14,1782 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...