Norbornadiene 97% stabilized

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 250 ppm

L13935-AC L13935-AE
CAAAAL13935-ACEA 87.37 CAD
CAAAAL13935-AC CAAAAL13935-AE
Norbornadiene 97% stabilized
Norbornadiene

Purity: 97% CAS No.: 121-46-0 Molecular Formula: C7H8 Formula Weight: 92.14 Flash Point: -11°(12°F) Refractive Index: 1.4700 Melting Point: -20° Boiling Point: 88-90° Beilstein Reference: 506224 UN No.: 2251 MDL No.: MFCD00082301

Formula: C₇H₈
Boiling Pt: 88…90 °C
Melting Pt: –20 °C
Density: 0.906
Flash Pt: –11 °C (12 °F)
Storage Temperature: Ambient
MDL Number: MFCD00082301
CAS Number: 121-46-0
EINECS: 204-472-0
UN: 2251
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR