L-α-Aminoadipic acid 98%
Supplier: Thermo Scientific Chemicals
L13924-03
L13924-06
CAAAAL13924-03EA
157.05
CAD
CAAAAL13924-03
CAAAAL13924-06
L-α-Aminoadipic acid 98%
L-α-Aminoadipic acid
Purity: 98%
CAS No.: 1118-90-7
Molecular Formula: C6H11NO4
Formula Weight: 161.16
Melting Point: ca 206° dec.
Beilstein Reference: 1724348
Formula:
C₆H₁₁NO₄ Melting Pt: ca 206 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002636 CAS Number: 1118-90-7 |
Specification Test Results
Optical rotation | +25° (c=2 in 5N HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...