Isopentyl nitrite (mixed isomers ) 97% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with sodium carbonate (Na₂CO₃) 0.2% (anhy)
L13909-AC
L13909-AE
L13909-AP
CAAAAL13909-ACEA
51.03
CAD
CAAAAL13909-AC
CAAAAL13909-AE
CAAAAL13909-AP
Isopentyl nitrite (mixed isomers ) 97% stabilized
Isopentyl nitrite (mixed isomers )
Fieser: 1,40 4,270 5,358 6,307 9,258
Purity: 97%
CAS No.: 110-46-3
Molecular Formula: (CH3)2CHCH2CH2ONO
Formula Weight: 117.15
Storage Temperature: 2°C to 8°C
Flash Point: -20°(-4°F)
Refractive Index: 1.3870
Odor: Sweet
Boiling Point: 95-100°
Beilstein Reference: 969510
Merck Reference: 14,5123
UN No.: 1113
MDL No.: MFCD00002057
Formula:
C₅H₁₁NO₂ Boiling Pt: 95…100 °C Density: 0.875 Flash Pt: -20 °C(-4 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002057 CAS Number: 110-46-3 EINECS: 203-770-8 UN: 1113 ADR: "3",II Merck Index: 14,5123 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...