L-(-)-tert-Leucine (H-Tle-OH) 99%

Supplier: Thermo Scientific Chemicals

L13707-03 L13707-06 L13707-14
CAAAAL13707-03EA 80.58 CAD
CAAAAL13707-03 CAAAAL13707-06 CAAAAL13707-14
L-(-)-tert-Leucine (H-Tle-OH) 99%
L(-)-tert-Leucine (H-Tle-OH)

Purity: 99% CAS No.: 20859-02-3 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Melting Point: 300° Beilstein Reference: 1721824

Formula: C₆H₁₃NO₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00064218
CAS Number: 20859-02-3
EINECS: 424-750-7

Order Now

Specification Test Results

Optical rotation +7° (c=5 in 5N HCl)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR