2-Dimethylaminoethyl methacrylate 97% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
DMA, N,N-Dimethylaminoethyl methacrylate
Stabilization:
Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ) ∼0.2%
L13507-AE
L13507-AP
CAAAAL13507-AEEA
59.41
CAD
CAAAAL13507-AE
CAAAAL13507-AP
2-Dimethylaminoethyl methacrylate 97% stabilized
2-Dimethylaminoethyl methacrylate
Purity: 97%
CAS No.: 2867-47-2
Molecular Formula: C8H15NO2
Formula Weight: 157.21
Storage Temperature: 2°C to 8°C
Flash Point: 70°(158°F)
Refractive Index: 1.4400
Melting Point: ca -30°
Boiling Point: 186-188°
Beilstein Reference: 1757048
UN No.: 2522
MDL No.: MFCD00008589
Formula:
C₈H₁₅NO₂ MW: 157.21 g/mol Boiling Pt: 186…188 °C Melting Pt: ca …30 °C Density: 0.932 Flash Pt: 70 °C(158 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008589 CAS Number: 2867-47-2 EINECS: 220-688-8 UN: 2522 ADR: "6.1",II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...