(4-Nitrophenyl)-α-D-galactopyranoside 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
(2R,3R,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol, 4-Nitrophenyl α-D-galactopyranoside
L13376-MC
L13376-ME
CAAAAL13376-MCEA
51.82
CAD
CAAAAL13376-MC
CAAAAL13376-ME
(4-Nitrophenyl)-α-D-galactopyranoside 98+%
(4-Nitrophenyl) α-D-galactopyranoside
Purity: 98+%
CAS No.: 7493-95-0
Molecular Formula: C12H15NO8
Formula Weight: 301.26
Storage Temperature: 2°C to 8°C
Melting Point: 166-169°
Beilstein Reference: 92214
Optical Rotation: +242° (c=1 in water)
MDL No.: MFCD00065050
Formula:
C₁₂H₁₅NO₈ MW: 301.26 g/mol Melting Pt: 166…169 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00065050 CAS Number: 7493-95-0 EINECS: 231-343-6 |
Specification Test Results
Optical rotation | +242° (c=1 in water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...