Sodium bis(trimethylsilyl)amide 2 M in THF

Supplier: Thermo Scientific Chemicals
Danger

L13352-AE L13352-AP
CAAAAL13352-AEEA 117.08 CAD
CAAAAL13352-AE CAAAAL13352-AP
Sodium bis(trimethylsilyl)amide 2 M in THF
Sodium bis(trimethylsilyl)amide

Fieser: 1,1046 3,261 4,442 6,529 7,329 12,441 18,332 20,349 21,401


CAS No.: 1070-89-9 Molecular Formula: C6H18NNaSi2 Formula Weight: 183.38 Flash Point: -17°(1°F) Beilstein Reference: 3629917 UN No.: 2924 MDL No.: MFCD00009835

Formula: C₆H₁₈NNaSi₂
Density: 0.920
Flash Pt: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009835
CAS Number: 1070-89-9
EINECS: 213-983-8
UN: 2924
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR