(S)-(+)-β-Aminophenethyl alcohol 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(S)-(+)-2-Phenylglycinol, L-Phenylglycinol, (S)-2-Amino-2-phenylethanol
L13265-03
L13265-06
L13265-14
CAAAAL13265-03EA
68.41
CAD
CAAAAL13265-03
CAAAAL13265-06
CAAAAL13265-14
(S)-(+)-β-Aminophenethyl alcohol 98+%
(S)-(+)-β-Aminophenethyl alcohol
Fieser: 15,256
Purity: 98+%
CAS No.: 20989-17-7
Molecular Formula: C8H11NO
Formula Weight: 137.18
Melting Point: 76-78°
Beilstein Reference: 3196190
Formula:
C₈H₁₁NO Melting Pt: 76…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064404 CAS Number: 20989-17-7 UN: 3263 ADR: "8",III |
Specification Test Results
Optical rotation | +30° (c=0.75 in 1N HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...