Ethylene glycol monoisopropyl ether 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L13241-0F
L13241-AE
L13241-AU
CAAAAL13241-0FEA
140.78
CAD
CAAAAL13241-0F
CAAAAL13241-AE
CAAAAL13241-AU
Ethylene glycol monoisopropyl ether 99%
Ethylene glycol monoisopropyl ether
Purity: 99%
CAS No.: 109-59-1
Molecular Formula: C5H12O2
Formula Weight: 104.15
Flash Point: 45°(113°F)
Refractive Index: 1.4100
Melting Point: -60°
Boiling Point: 143°
Beilstein Reference: 1732184
UN No.: 1992
MDL No.: MFCD00002866
Formula:
C₅H₁₂O₂ Boiling Pt: 143 °C Melting Pt: –60 °C Density: 0.903 Flash Pt: 45 °C(113 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002866 CAS Number: 109-59-1 EINECS: 203-685-6 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...